When versatility, low limits of detection and high sample throughput are critical, industrial businesses rely on Thermo Scientific™ Niton™ handheld XRF analyzers. Providing customers with solutions designed to meet their most demanding applications, Niton analyzers are built to maximize performance and productivity. Our comprehensive line of portable XRF analyzers have been created to meet your unique challenges and budget.
Discover how thousands of customers have generated rapid quantitative and qualitative results in almost every location, field environment and weather condition. Applications include, metal alloy analysis, oil and gas exploration, mining and geology examination, toys and consumer goods testing and environmental analysis/remediation. Read more about our versatile, easy-to-use XRF guns below.
Thermo Scientific™ Niton XL5
Premium Offering
Powered by a 5W x-ray tube and weighing an industry leading 2.8 lbs
(1.3 kg), the Niton XL5 is small but mighty. A miniaturized footprint means you'll reach tight spaces with ease, while reducing operator fatigue. An application interface and WiFi accessibility provide the latest in technology features, while a micro and macro camera enable improved record keeping.
Thermo Scientific™ Niton XL3
Standard Solution
The Niton XL3 delivers a feature rich package built to help you obtain rapid identification of up to 30 elements ranging from Mg to U. Easily view elemental makeup and confidence on the intuitive results screen. Point and shoot simplicity ensures that non-technical users are able to operate with minimal instruction.
Thermo Scientific™ Niton XL2
Value Leader
Our tried and true offering, the Niton XL2 is recognized for its performance and durability. Obtain fast and accurate elemental analysis of metals, alloys, precious metals, geological samples and environmental contaminants. With the Niton XL2, you don't have to sacrifice performance for analytical power.
Free eBook Download
Interested in learning how handheld XRF analysis actually works? Download the free eBook, "XRF Technology for Non-Scientists: XRF Technology In the Field."